Thermo Scientific Chemicals

Brefeldin A, 98%, Thermo Scientific Chemicals

Catalog number: J62340.MF
50 mg, Each
Thermo Scientific Chemicals

Brefeldin A, 98%, Thermo Scientific Chemicals

Catalog number: J62340.MF
50 mg, Each
Quantity
Catalog number: J62340.MF
also known as J62340-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20350-15-6
IUPAC Name
(1S,6S,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-1H,4H,6H,7H,8H,9H,11aH,12H,13H,14H,14aH-cyclopenta[f]oxacyclotridecan-4-one
Molecular Formula
C16H24O4
InChI Key
KQNZDYYTLMIZCT-PNFJWZTBSA-N
SMILES
C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1
Form
Crystals or powder or crystalline powder or granules
Appearance (Color)
White to pale yellow
Assay (HPLC)
≥97.5%

Description

Brefeldin A mediated apoptosis has been observed in human tumor cellsIt is an antibiotic used in cell culture and to inhibit protein secretion in mammalian and other eukaryotic cells by interfering with the function of the Golgi apparatus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brefeldin A mediated apoptosis has been observed in human tumor cellsIt is an antibiotic used in cell culture and to inhibit protein secretion in mammalian and other eukaryotic cells by interfering with the function of the Golgi apparatus.

Solubility
Soluble in dimethylsulfoxide, dichloromethane and ethanol. Slightly soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text