Thermo Scientific Chemicals

Trichloroacetic acid, 5% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: J62366.AP
500 mL, Each
Thermo Scientific Chemicals

Trichloroacetic acid, 5% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: J62366.AP
500 mL, Each
Quantity
Catalog number: J62366.AP
also known as J62366-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-03-9
IUPAC Name
trichloroacetic acid
Molecular Formula
C2HCl3O2
InChI Key
YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES
OC(=O)C(Cl)(Cl)Cl
Appearance (Color)
Clear colorless
Form
Liquid
Assay from Supplier's CofA
4.75-5.25%

Description

Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.

Solubility
Soluble in water (1200 mg/ml at 20°C), alcohol, ether, ethanol (very soluble), and carbon tetrachloride.

Notes
Store under nitrogen. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases, smines.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text