Thermo Scientific Chemicals

Irinotecan hydrochloride trihydrate, 99%, Thermo Scientific Chemicals

Catalog number: J62370.MC
100 mg, Each
Thermo Scientific Chemicals

Irinotecan hydrochloride trihydrate, 99%, Thermo Scientific Chemicals

Catalog number: J62370.MC
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62370.MC
also known as J62370-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
136572-09-3
IUPAC Name
hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate trihydrate chloride
Molecular Formula
C33H45ClN4O9
InChI Key
KLEAIHJJLUAXIQ-JDRGBKBRSA-N
SMILES
[H+].O.O.O.[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O
Assay (HPLC)
> 98.5%

Description

Topoisomerase I inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text