Thermo Scientific Chemicals

Potassium phosphate, 0.2M buffer soln., pH 7.4

Catalog number: J62397.K2
1 L, Each
Thermo Scientific Chemicals

Potassium phosphate, 0.2M buffer soln., pH 7.4

Catalog number: J62397.K2
1 L, Each
Quantity
Catalog number: J62397.K2
also known as J62397-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7778-77-0
IUPAC Name
potassium dihydrogen phosphate
Molecular Formula
H2KO4P
InChI Key
GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES
[K+].OP(O)([O-])=O
Appearance (Color)
Clear colorless
Form
Liquid
Concentration
Potassium Phosphate Monobasic/Potassium Phosphate Dibasic: 200 mM
pH
7.2-7.6

Description

Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.

Solubility
Soluble in water, Insoluble to slightly soluble in ethanol, insoluble in alcohol.

Notes
Store sealed bottle containing the dehydrated medium at 2 - 30C. Once opened and recapped, place container in a low humidity environment at the same storage temperature. Protect from moisture and light by keeping container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text