Thermo Scientific Chemicals

Glycine, 99+%, Cell Culture Reagent, Thermo Scientific Chemicals

Catalog number: J62407.36
500 g, Each
Thermo Scientific Chemicals

Glycine, 99+%, Cell Culture Reagent, Thermo Scientific Chemicals

Catalog number: J62407.36
500 g, Each
Quantity
Catalog number: J62407.36
also known as J62407-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-40-6
IUPAC Name
2-aminoacetic acid
Molecular Formula
C2H5NO2
InChI Key
DHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILES
NCC(O)=O
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Assay from Suppliers CofA
≥99.0% (unspecified)
Loss on Drying
≤0.2%

Description

This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
Soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature 2 - 8°C
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text