Denhardt's solution (100X)
This product is animal origin and can only be shipped within the United States
Denhardt's solution (100X)
Thermo Scientific Chemicals

Denhardt's solution (100X)

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62416.AD
also known as J62416-AD
Price (USD)/ Each
137.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
162.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
137.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
162.00 
Save 24.35 (15%)
Add to cart
Denhardt's solution (100X)
Catalog numberJ62416.AD
Price (USD)/ Each
137.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
162.00 
Save 24.35 (15%)
-
Add to cart
Specifications
Chemical Name or MaterialDenhardt's solution
Recommended StorageStore at -20°C
TSCAYes
MDL NumberMFCD00717549
Concentration100X
View more
Denhardt's Solution is used as a blocking reagent for northern or southern blots to prevent probe binding on open membrane space and non-target nucleic acids. Denhardt's reagent is recommended for Northern hybridization, hybridization of RNA, single copy Southern hybridization' or hybridization of DNA, immobilized on nylon membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only