Polyvinylpyrrolidone, M.W. 40,000, Thermo Scientific Chemicals
Polyvinylpyrrolidone, M.W. 40,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyvinylpyrrolidone, M.W. 40,000, Thermo Scientific Chemicals

A cryoprotectant, also useful in plant cell culture research
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Catalog number J62417.36
also known as J62417-36
Price (USD)/ Each
117.65
Online exclusive
131.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
117.65
Online exclusive
131.00 
Save 13.35 (10%)
Add to cart
Polyvinylpyrrolidone, M.W. 40,000, Thermo Scientific Chemicals
Catalog numberJ62417.36
Price (USD)/ Each
117.65
Online exclusive
131.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Elemental AnalysisNitrogen: 11.5-12.8%
Water Content (Karl Fischer Titration)≤5%
pH3.0-7.0
Assay (special)K value: 27-32
View more
A cryoprotectant, also useful in plant cell culture research Polyvinylpyrrolidone is used as an adhesive in glue sticks; an emulsifier and a disintegrant for solution polymerization; an additive to Doro's RNA extraction buffer; as a liquid-phase dispersion enhancing agent in diffusion-ordered spectroscopy (DOSY) NMR and as a thickening agent in tooth whitening gels. It finds use in personal care products like shampoos and toothpastes, in ink for inkjet printers as well as in contact lens solutions. It is used as a food additive and in the wine industry as a fining agent for white wine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only