Oxytetracycline, 96%, Thermo Scientific Chemicals
Oxytetracycline, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Oxytetracycline, 96%, Thermo Scientific Chemicals

Oxytetracycline, CAS # 79-57-2, shows antibacterial, anti-inflammatory, and anti-proliferative activities. It can also inhibit mitochondrial protein synthesis.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62427.18
also known as J62427-18
Price (USD)/ Each
184.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
217.00 
Save 32.35 (15%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
184.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
217.00 
Save 32.35 (15%)
Add to cart
Oxytetracycline, 96%, Thermo Scientific Chemicals
Catalog numberJ62427.18
Price (USD)/ Each
184.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
217.00 
Save 32.35 (15%)
-
Add to cart
Chemical Identifiers
CAS79-57-2
IUPAC Namedisodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate
Molecular FormulaC22H26N2Na2O11
InChI KeyOLRBUPVCLOPDOT-GETZKYKJSA-M
SMILESO.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)C(=C(N)O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (special)Identificaion: Positive
Appearance (Color)Yellow
pH4.5 to 7.0
Assay (HPLC)≥ 96.0%
Oxytetracycline is an anti-inflammatory, antibiotic, and anti-proliferative agent shown to inhibit mitochondrial protein synthesis. Mechanistic studies show that this compound reduces cytochrome c activity which eventually causes cell growth arrest. Additionally, Oxytetracycline has demonstrated the ability to reduce murine bronchial inflammation. The anti-inflammatory characteristics of this agent are due to its ability to cause a decrease in IL-4, IL-5, IL-13, immune cells, and the ovalbumin-specific IgE. Data suggests that Oxytetracycline is also an inhibitor of NF-κB activation via phosphorylation and degradation of IκB-α.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Oxytetracycline is a compound with broad-spectrum antibacterial activity. Furthermore, it also shows anti-inflammatory and anti-proliferative activities
  • It can inhibit mitochondrial protein synthesis in both gram-positive and gram-negative bacteria

Application

  • Oxytetracycline is active against several species including Mycoplasma pneumonia, Pasteurella pestis, Escherichia coli, Haemophilus influenza, and Diplococcus pneumonia
  • In laboratory experiments, oxytetracycline can be used to evaluate the acquisition of oxytetracycline-resistance genes, which are associated with the development of antibiotic resistance
  • It can reduce cytochrome c activity, causing cell growth arrest
  • It has been shown to reduce murine bronchial inflammation
  • Oxytetracycline exerts its anti-inflammatory activity by decreasing the levels of IL-4, IL-5, IL-13 immune cells, and the Ovalbumin-specific IgE
  • Oxytetracycline can be used to inhibit NF-κB activation via phosphorylation and degradation of IκB-α
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Harry Björklund.; Johan Bondestam.; Göran Bylund. Residues of oxytetracycline in wild fish and sediments from fish farms.Aquaculture. 1990, 86 (4),359-367 .
  2. Ole Bent Samuelsen.; Vigdis Torsvik.; Arne Ervik. Long-range changes in oxytetracycline concentration and bacterial resistance towards oxytetracycline in a fish farm sediment after medication.Sci. Total Environ. 1992, 114 25-36 .