Thermo Scientific Chemicals

Oxytetracycline, 96%, Thermo Scientific Chemicals

Catalog number: J62427.22
100 g, Each
Thermo Scientific Chemicals

Oxytetracycline, 96%, Thermo Scientific Chemicals

Catalog number: J62427.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62427.22
also known as J62427-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-57-2
IUPAC Name
disodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate
Molecular Formula
C22H26N2Na2O11
InChI Key
OLRBUPVCLOPDOT-GETZKYKJSA-M
SMILES
O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)C(=C(N)O)C1=O
Form
Crystalline powder
Assay (special)
Identificaion: Positive
Appearance (Color)
Yellow
pH
4.5 to 7.0
Assay (HPLC)
≥ 96.0%

Description

Oxytetracycline is an anti-inflammatory, antibiotic, and anti-proliferative agent shown to inhibit mitochondrial protein synthesis. Mechanistic studies show that this compound reduces cytochrome c activity which eventually causes cell growth arrest. Additionally, Oxytetracycline has demonstrated the ability to reduce murine bronchial inflammation. The anti-inflammatory characteristics of this agent are due to its ability to cause a decrease in IL-4, IL-5, IL-13, immune cells, and the ovalbumin-specific IgE. Data suggests that Oxytetracycline is also an inhibitor of NF-κB activation via phosphorylation and degradation of IκB-α.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Oxytetracycline is a compound with broad-spectrum antibacterial activity. Furthermore, it also shows anti-inflammatory and anti-proliferative activities
  • It can inhibit mitochondrial protein synthesis in both gram-positive and gram-negative bacteria

Application

  • Oxytetracycline is active against several species including Mycoplasma pneumonia, Pasteurella pestis, Escherichia coli, Haemophilus influenza, and Diplococcus pneumonia
  • In laboratory experiments, oxytetracycline can be used to evaluate the acquisition of oxytetracycline-resistance genes, which are associated with the development of antibiotic resistance
  • It can reduce cytochrome c activity, causing cell growth arrest
  • It has been shown to reduce murine bronchial inflammation
  • Oxytetracycline exerts its anti-inflammatory activity by decreasing the levels of IL-4, IL-5, IL-13 immune cells, and the Ovalbumin-specific IgE
  • Oxytetracycline can be used to inhibit NF-κB activation via phosphorylation and degradation of IκB-α
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text