Penicillin V potassium salt
Penicillin V potassium salt
Penicillin V potassium salt
Penicillin V potassium salt
Thermo Scientific Chemicals

Penicillin V potassium salt

Inhibits bacterial cell wall synthesis | CAS: 132-98-9 | C16H17KN2O5S | 388.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62442.09
also known as J62442-09
Price (USD)
68.40
Each
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
68.40
Each
Add to cart
Chemical Identifiers
CAS132-98-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥98.0%
Mass SpecConsistent with structure
Ask our AI about this Product
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.

Solubility
Soluble in water, dimethyl sulfoxide, and methanol.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
RUO – Research Use Only

General References:

  1. Li-Li Du, Qi Wu.; Chun-Xiu Chen.; Bo-Kai Liu.; Xian-Fu Lin. A two-step, one-pot enzymatic synthesis of ampicillin from penicillin G potassium salt.J. Mol. Catal. B: Enzym. 2009, 58 (1-4),208-211 .
  2. John C. Sheehan.; Kenneth R. Henery-Logan. The Total Synthesis of Penicillin V.J. Am. Chem. Soc. 1959, 81 (12),3089-3094 .
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link