Thermo Scientific Chemicals

TBE (10X), TRIS + borate + EDTA

Catalog number: J62449.K2
1 L, Each
Thermo Scientific Chemicals

TBE (10X), TRIS + borate + EDTA

Catalog number: J62449.K2
1 L, Each
Quantity
Catalog number: J62449.K2
also known as J62449-K2
Price (USD)
Quantity
-
Have Questions?
Form
Liquid
Composition
EDTA: 20mM
Comment
Filtered through 0.22 micron filter
Appearance (Color)
Clear colorless
Composition
Boric acid: 890mM

Description

TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. TBE Buffer is used for polyacrylamide and agarose gel electrophoresis. It is ideally suited for DNA and RNA separation on longer run, higher voltage or amperage gels. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel.5 TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids. Tris-Borate-EDTA buffer has been used for pulsed-field gel electrophoresis (PFGE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text