1x1 image pixel for data collection
Okadaic acid sodium salt, 98%, Thermo Scientific Chemicals
Okadaic acid sodium salt, 98%, Thermo Scientific Chemicals
Okadaic acid sodium salt, 98%, Thermo Scientific Chemicals
Okadaic acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Okadaic acid sodium salt, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 μg
100 μg
300 μg
Catalog number J62463.IAM
also known as J62463-I&
Price (USD)
99.65
Special offer
Online exclusive
Ends: 26-Sep-2025
123.00
Save 23.35 (19%)
Each
Add to cart
Quantity:
50 μg
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 26-Sep-2025
123.00
Save 23.35 (19%)
Each
Add to cart
Specifications
CAS209266-80-8
Chemical Name or MaterialOkadaic acid sodium salt
ColorWhite
DOT InformationHazard Class: 6.1; Packaging Group: II
Formula Weight827
View more
Potent inhibitor of serine/threonine-specific protein phosphatases 1 and 2A (PP1 and PP2A). Does not inhibit tyrosine or alkaline phosphatases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Potent inhibitor of serine/threonine-specific protein phosphatases 1 and 2A (PP1 and PP2A)
  • Does not inhibit tyrosine or alkaline phosphatases
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link