Thermo Scientific Chemicals

Rapamycin, 98+%, Thermo Scientific Chemicals

Catalog number: J62473.EX3
200 mg, Each
Thermo Scientific Chemicals

Rapamycin, 98+%, Thermo Scientific Chemicals

Catalog number: J62473.EX3
200 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62473.EX3
also known as J62473-!3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53123-88-9
IUPAC Name
1,18-dihydroxy-12-[1-(4-hydroxy-3-methoxycyclohexyl)propan-2-yl]-19,30-dimethoxy-15,17,21,23,29,35-hexamethyl-11,36-dioxa-4-azatricyclo[30.3.1.0⁴,⁹]hexatriaconta-16,24,26,28-tetraene-2,3,10,14,20-pentone
Molecular Formula
C51H79NO13
InChI Key
QFJCIRLUMZQUOT-UHFFFAOYNA-N
SMILES
COC1CC(CC(C)C2CC(=O)C(C)C=C(C)C(O)C(OC)C(=O)C(C)CC(C)C=CC=CC=C(C)C(CC3CCC(C)C(O)(O3)C(=O)C(=O)N3CCCCC3C(=O)O2)OC)CCC1O
Appearance (Color)
White to cream to yellow
Form
Powder or crystals or crystalline powder
Assay from Suppliers CofA
≥98.0%

Description

Blocks cytokine-mediated signal transduction pathwaysRapamycin is used to inhibit tumor growth by halting tumor cell proliferation and suppressing tumor angiogenesis. It forms a complex with FKBP12 that binds and inhibits the molecular target of rapamycin (mTOR) as well as suppresses the immune system, preventing transplant rejection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Rapamycin is a natural macrolide compound showing antiproliferative activities in mammalian cells
  • Rapamycin is a specific inhibitor of rapamycin kinase. It forms a complex with FKBP12 that consequently binds and inhibits rapamycin’s molecular target (rapamycin kinase)

Application

  • Rapamycin inhibits tumor growth by preventing tumor cell proliferation and suppressing tumor angiogenesis
  • This compound also blocks cytokine-mediated signal transduction pathways
  • Rapamycin also promotes the suppression of the immune system
  • It was initially defined as an antifungal metabolite produced by Streptomyces hygroscopicus
  • In vitro studies demonstrated that this compound is an important mediator of PI3 kinase signaling
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text