Thermo Scientific Chemicals

Camptothecin

Catalog number: J62523.03
1 g, Each
Thermo Scientific Chemicals

Camptothecin

Catalog number: J62523.03
1 g, Each
Quantity
Catalog number: J62523.03
also known as J62523-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7689-03-4
IUPAC Name
(19S)-19-ethyl-19-hydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Molecular Formula
C20H16N2O4
InChI Key
VSJKWCGYPAHWDS-FQEVSTJZSA-N
SMILES
CC[C@@]1(O)C(=O)OCC2=C1C=C1N(CC3=C1N=C1C=CC=CC1=C3)C2=O
Assay (HPLC)
>90%

Description

Camptothecin is used as a cytotoxic antitumor agent that inhibits topoisomerase I.Camptothecin shows remarkable anticancer activity based on preliminary clinical trials. Its analogues are used in cancer chemotherapy. Cyclodextrin-based polymer containing camptothecin is used in the investigational anti-cancer drug study.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text