Camptothecin
Thermo Scientific Chemicals

Camptothecin

Cytotoxic antitumor agent that inhibits topoisomerase I
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number J62523.MD
also known as J62523-MD
Price (USD)/ Each
87.65
Online exclusive
97.30 
Save 9.65 (10%)
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
87.65
Online exclusive
97.30 
Save 9.65 (10%)
Add to cart
Camptothecin
Catalog numberJ62523.MD
Price (USD)/ Each
87.65
Online exclusive
97.30 
Save 9.65 (10%)
-
Add to cart
Chemical Identifiers
CAS7689-03-4
IUPAC Name(19S)-19-ethyl-19-hydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Molecular FormulaC20H16N2O4
InChI KeyVSJKWCGYPAHWDS-FQEVSTJZSA-N
SMILESCC[C@@]1(O)C(=O)OCC2=C1C=C1N(CC3=C1N=C1C=CC=CC1=C3)C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>90%
Camptothecin is used as a cytotoxic antitumor agent that inhibits topoisomerase I.Camptothecin shows remarkable anticancer activity based on preliminary clinical trials. Its analogues are used in cancer chemotherapy. Cyclodextrin-based polymer containing camptothecin is used in the investigational anti-cancer drug study.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C