Thermo Scientific Chemicals

Glycine, 0.2M buffer soln., pH 3.0

Catalog number: J62527.AP
500 mL, Each
Thermo Scientific Chemicals

Glycine, 0.2M buffer soln., pH 3.0

Catalog number: J62527.AP
500 mL, Each
Quantity
Catalog number: J62527.AP
also known as J62527-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-40-6
IUPAC Name
2-aminoacetic acid
Molecular Formula
C2H5NO2
InChI Key
DHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILES
NCC(O)=O
Appearance (Color)
Clear colorless
Form
Liquid
Comment
Component is dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and filtered through 1 micron filter.
pH
3.0 ± 0.2

Description

Glycine is widely used as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine is widely used as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
Not miscible or difficult to mix in water. Slightly soluble in acetone. Insoluble in alcohol, and ether.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text