Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 10X), with 1% Triton™ X-100, Thermo Scientific Chemicals

Catalog number: J62533.AP
500 mL, Each
Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 10X), with 1% Triton™ X-100, Thermo Scientific Chemicals

Catalog number: J62533.AP
500 mL, Each
Quantity
Catalog number: J62533.AP
also known as J62533-AP
Price (USD)
Quantity
-
Have Questions?

Description

It used in some biochemical techniques that is maintain the pH within a relatively narrow range. Tris-HCl is commonly used to make TBS buffers. Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash micro titer plates for western blot experiments and screening methods. it is suitable for use as a washing buffer for alkaline phosphatase or peroxidase conjugates in dot blot assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It used in some biochemical techniques that is maintain the pH within a relatively narrow range. Tris-HCl is commonly used to make TBS buffers. Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash micro titer plates for western blot experiments and screening methods. it is suitable for use as a washing buffer for alkaline phosphatase or peroxidase conjugates in dot blot assay.

Solubility
Fully miscible in water.

Notes
250mM Tris, 27mM KCl, 1.37M NaCl, pH 7.4
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text