Thermo Scientific Chemicals
Nitrocellulose stripping buffer (10X)
Catalog number: J62541.AK
250 mL, Each
Thermo Scientific Chemicals
Nitrocellulose stripping buffer (10X)
Catalog number: J62541.AK
250 mL, Each
Quantity
Catalog number: J62541.AK
also known as J62541-AK
Price (USD)
Quantity
-
Have Questions?
Description
Ideal for removal of secondary antibody after detection of highly abundant targets. Nitrocellulose Stripping Buffer may perform well with nitrocellulose membranes or antibody-dye conjugates. It is also employed for nucleic acid and protein related blotting works.