beta-Nicotinamide adenine dinucleotide phosphate disodium salt, 97%, Thermo Scientific Chemicals
beta-Nicotinamide adenine dinucleotide phosphate disodium salt, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

beta-Nicotinamide adenine dinucleotide phosphate disodium salt, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number J62556.03
also known as J62556-03
Price (USD)/ Each
343.65
Online exclusive
382.00 
Save 38.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
343.65
Online exclusive
382.00 
Save 38.35 (10%)
Add to cart
beta-Nicotinamide adenine dinucleotide phosphate disodium salt, 97%, Thermo Scientific Chemicals
Catalog numberJ62556.03
Price (USD)/ Each
343.65
Online exclusive
382.00 
Save 38.35 (10%)
-
Add to cart
Chemical Identifiers
CAS24292-60-2
IUPAC Namedisodium 1-[(2R,3R,4S,5R)-5-({[({[(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-(hydrogen phosphonatooxy)-3-hydroxyoxolan-2-yl]methyl phosphonato}oxy)(oxido)phosphoryl]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Molecular FormulaC21H26N7Na2O17P3
InChI KeyWSDDJLMGYRLUKR-MWLGWILCNA-L
SMILES[Na+].[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale yellow
FormCrystalline powder
Assay (special)? 97% (dry weight basis)
Heavy Metals (as Pb)? 5 ppm
Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications, especially anaerobic reactions such as lipid and nucleic acid synthesis. NADP/NADPH is a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems, such as the thioredoxin reductase/thioredoxin system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications, especially anaerobic reactions such as lipid and nucleic acid synthesis. NADP/NADPH is a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems, such as the thioredoxin reductase/thioredoxin system.

Solubility
Soluble in water (50 mg/ml), 0.001% aqueous buffer, pH 7.0, methanol, and ethanol (less soluble). Insoluble in ether, and ethyl acetate.

Notes
Protect from light and moisture. Keep cool and dry. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Store under inert gas.
RUO – Research Use Only

General References:

  1. PR Coulet.; JH Julliard. A mild method of general use for covalent coupling of enzymes to chemically activated collagen films. Biotechnology and Bioengineering. 1974, 16, (8), 1055-1068.
  2. H Somer.; A Konttinen. Demonstration of serum creatine kinase isoenzymes by fluorescence technique. Clinica Chimica Acta. 1972, 40, (1), 133-138.