Carbamazepine, 98%, Thermo Scientific Chemicals
Carbamazepine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Carbamazepine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62590.14
also known as J62590-14
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
Add to cart
Carbamazepine, 98%, Thermo Scientific Chemicals
Catalog numberJ62590.14
Price (USD)/ Each
116.65
Online exclusive
129.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS298-46-4
IUPAC Name2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide
Molecular FormulaC15H12N2O
InChI KeyFFGPTBGBLSHEPO-UHFFFAOYSA-N
SMILESNC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay from Suppliers CofA≥97.5%
FormPowder
Carbamazepine is used as cytochrome P450 3A4 inducing anti-epileptic drug and ligand for the GABA-A receptor benzodiazepine modulatory site. Carbamazepine is typically used for the treatment of seizure disorders and neuropathic pain. It is associated with other medications and used to treat schizophrenia as well as a second line agent in bipolar disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol, acetone, propylene glycol, chloroform, dimethyl sulfoxide, methanol, dimethyl fluoride,ethylene glycol and monoethyl ether. Slightly soluble in glacial acetic acid. Insoluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Tolbert, D.; Cloyd, J.; Biton, V.; Bekersky, I.; Walzer, M.; Wesche, D.; Drummond, R.; Lee, D. Bioequivalence of oral and intravenous carbamazepine formulations in adult patients with epilepsy. Epilepsia. 2015, 56 (6), 915-923.
  2. Shokry, E.; Villanelli, F.; Malvagia, S.; Rosati, A.; Forni, G.; Funghini, S.; Ombrone, D.; Bona, M. D.; Guerrini, R.; Marca, G. L. Therapeutic drug monitoring of carbamazepine and its metabolite in children from dried blood spots using liquid chromatography and tandem mass spectrometry. J. Pharm. Biomed. Anal. 2015, 109, 164-170.