Imidazole, 0.5M buffer soln., pH 7.0
Imidazole, 0.5M buffer soln., pH 7.0
Thermo Scientific Chemicals

Imidazole, 0.5M buffer soln., pH 7.0

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J62593.AE
also known as J62593-AE
Price (USD)/ Each
90.60
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
90.60
Add to cart
Imidazole, 0.5M buffer soln., pH 7.0
Catalog numberJ62593.AE
Price (USD)/ Each
90.60
-
Add to cart
Specifications
Concentration0.5M buffer soln.
Molecular Weight (g/mol)68.079
pH7.0
Molecular FormulaC3H4N2
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Keep cold