Tylosin tartrate, 94%, Thermo Scientific Chemicals
Tylosin tartrate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tylosin tartrate, 94%, Thermo Scientific Chemicals

Macrolide antibiotic
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62633.06
also known as J62633-06
Price (USD)/ Each
176.65
Online exclusive
196.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
176.65
Online exclusive
196.00 
Save 19.35 (10%)
Add to cart
Tylosin tartrate, 94%, Thermo Scientific Chemicals
Catalog numberJ62633.06
Price (USD)/ Each
176.65
Online exclusive
196.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1405-54-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow
Assay (unspecified)≥92.0%
Assay (unspecified)Tyramine: ≤0.35%
Loss on Drying≤4.5%
Sulfated Ash≤2.5%
View more
It is used as an antibacterial agent. This compound has been shown to be effective in vitro against Staphylococcus intermedius and sensitive mycoplasma organisms. Although a lot of antibiotics tend to suppress the immune system, macrolide antibiotics have been reported to enhance the immune system when used in chicken models. Studies suggest that chronic exposure to this compound may have a negative effect on the respiratory system. For turkeys - for treatment of infectious synovitis and sinusitis. It is effective also for treatment of ileitis, dysentery and necrotic enteritis in swine. It is successfully applied as a preventive measure in stress conditions as vaccinations, transportation and others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an antibacterial agent. This compound has been shown to be effective in vitro against Staphylococcus intermedius and sensitive mycoplasma organisms. Although a lot of antibiotics tend to suppress the immune system, macrolide antibiotics have been reported to enhance the immune system when used in chicken models. Studies suggest that chronic exposure to this compound may have a negative effect on the respiratory system. For turkeys - for treatment of infectious synovitis and sinusitis. It is effective also for treatment of ileitis, dysentery and necrotic enteritis in swine. It is successfully applied as a preventive measure in stress conditions as vaccinations, transportation and others.

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

General References:

  1. Knothe H. A review of the medical considerations of the use of tylosin and other macrolide antibiotics as additives in animal feeds.Infection.,1977,5(3), 183-7.
  2. Lee HS, et al. Occupational asthma due to tylosin tartrate.Br J Ind Med.,1989,46(7), 498-9.