Thermo Scientific Chemicals

Sorbitan sesquioleate, Thermo Scientific Chemicals

Catalog number: J62648.K2
1 L, Each
Thermo Scientific Chemicals

Sorbitan sesquioleate, Thermo Scientific Chemicals

Catalog number: J62648.K2
1 L, Each
Quantity
Catalog number: J62648.K2
also known as J62648-K2
Price (USD)
Quantity
-
Have Questions?

Description

Sorbitan sesquioleate is an additive found in food, veterinary products, and household items. Sorbitan esters being nonionic surface agents are used as excipients in veterinary medicinal products. Emollient ester of this compound offers a smooth feel 'coated' after-feel to the skin and is used in cosmetics. It acts as a emulsification and lubricating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan sesquioleate is an additive found in food, veterinary products, and household items. Sorbitan esters being nonionic surface agents are used as excipients in veterinary medicinal products. Emollient ester of this compound offers a smooth feel ‘coated‘ after-feel to the skin and is used in cosmetics. It acts as a emulsification and lubricating agent.

Solubility
Insoluble in cold water, hot water. soluble in mineral oil and ethyl acetate.

Notes
Keep container tightly closed. Keep container in a cool, well-ventilated area and protect from moisture. 
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text