Thermo Scientific Chemicals
TRIS-buffered saline (TBS, 10X) pH 7.6
Catalog number: J62662.K3
2 L, Each
Thermo Scientific Chemicals
TRIS-buffered saline (TBS, 10X) pH 7.6
Catalog number: J62662.K3
2 L, Each
Quantity
Catalog number: J62662.K3
also known as J62662-K3
Price (USD)
Quantity
-
Have Questions?
Description
TRIS-buffered saline (TBS, 10X) pH 7.6 is used as a substance diluent, as a wash buffer in immunoassays such as enzyme-linked immunosorbent assay. It is used in antibody diluent in western blotting, sample diluent in vitro diagnostics, immuno-histochemical staining and in situ hybridization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Miscible with water.
Notes
250mM Tris, 27mM potassium chloride, 1.37M sodium chloride, pH 7.6.Store in a cool place. Incompatible with strong oxidizing agents.