Thermo Scientific Chemicals

Tolmetin sodium salt dihydrate, 98+%, Thermo Scientific Chemicals

Catalog number: J62663.14
25 g, Each
Thermo Scientific Chemicals

Tolmetin sodium salt dihydrate, 98+%, Thermo Scientific Chemicals

Catalog number: J62663.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62663.14
also known as J62663-14
Price (USD)
Quantity
-
Have Questions?

Description

Tolmetin Sodium is an anti-inflammatory, antirheumatic agent. It can inhibit aldo-keto reductases (e.g., IC50 = 2.39 μM for human aldose reductase), a family of enzymes that catalyze the NADPH-dependent reduction of carbonyl groups in a number of important steroid, metabolic, and prostanoid molecules. It is a beta-adrenergic blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tolmetin Sodium is an anti-inflammatory, antirheumatic agent. It can inhibit aldo-keto reductases (e.g., IC50 = 2.39 µM for human aldose reductase), a family of enzymes that catalyze the NADPH-dependent reduction of carbonyl groups in a number of important steroid, metabolic, and prostanoid molecules. It is a beta-adrenergic blocker.

Solubility
Soluble in water

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text