General description
• G418 sulfate (geneticin) is an aminoglycoside antibiotic isolated from Micromonospora rhodorangea
• G418 is a bacteriostatic that prevents polypeptide formation by inhibiting the elongation step of polypeptide synthesis
Applications
• G418 sulfate can be used for selection of genetically engineered cells in both eukaryotes and prokaryotes
• Laboratory grade for applications such as biochemistry
Literature references
Arshad, H.; Patel, Z.; Mehrabian, M.; Bourkas, M.; Al-Azzawi, Z.; Schmitt-Ulms, G., & Watts, J. C. 2021. The aminoglycoside G418 hinders de novo prion infection in cultured cells. The Journal of biological chemistry, 297(3), 101073.
Ferguson, M. W.; Gerak, C.; Chow, C.; Rastelli, E. J.; Elmore, K. E.; Stahl, F.; Hosseini-Farahabadi, S.; Baradaran-Heravi, A.; Coltart, D. M., & Roberge, M. 2019. The antimalarial drug mefloquine enhances TP53 premature termination codon readthrough by aminoglycoside G418. PloS one, 14(5), e0216423.