Thermo Scientific Chemicals

Cefotaxime sodium salt, Thermo Scientific Chemicals

Catalog number: J62690.03
1 g, Each
Thermo Scientific Chemicals

Cefotaxime sodium salt, Thermo Scientific Chemicals

Catalog number: J62690.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62690.03
also known as J62690-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64485-93-4
IUPAC Name
sodium (6R,7R)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Formula
C16H16N5NaO7S2
InChI Key
AZZMGZXNTDTSME-JUZDKLSSSA-M
SMILES
[Na+].CON=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
Loss on Drying
≤ 3.0%
Optical Rotation
+58 to 64?
pH
4.5 - 6.5
Form
Powder
Assay (unspecified)
(dried basis): 91.6 - 96.4%

Description

Cefotaxime sodium salt acts as a beta-lactamase resistant antibiotic. It is used as an effective antibacterial against gram-negative bacteria, with the notable exception of pseudomonas and penicillin-resistant strains of streptococcus pneumoniae. It is used to treat infections of the bones, joints, skin, respiratory tract and blood stream.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• The cefotaxime sodium salt is a - lactamase considered a third-generation cephalosporin with antibiotic and bactericidal activities
• This compound exerts its biological activity by binding to penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall leading to their inactivation. The inactivation of these proteins affects the cross-linking of peptidoglycan chains required for bacterial cell wall strength and rigidity, resulting in the weakening of cell walls and cell lysis.

Applications

• It is an efficient antibacterial compound that acts against gram-negative bacteria, without activity against gram-positive bacteria such as pseudomonas and penicillin-resistant strains of streptococcus pneumonia
• It has been used for plant cell culture and human cell lines in vitro
• Cefotaxime sodium salt has been used in antibiotic susceptibility testing (AST) methods
• Cefotaxime sodium salt is also used to remove bacteria from Ostreococcus tauri cultures

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text