Phosphate-buffered saline (PBS, 10X), pH 7.6
Phosphate-buffered saline (PBS, 10X), pH 7.6
Thermo Scientific Chemicals

Phosphate-buffered saline (PBS, 10X), pH 7.6

Helps to maintain a constant pH
Have Questions?
Change viewbuttonViewtableView
Capacity:
1 L
4 L
2 L
Catalog number J62692.K2
also known as J62692-K2
Price (USD)/ Each
75.65
Online exclusive
84.40 
Save 8.75 (10%)
-
Add to cart
Capacity:
1 L
Request bulk or custom format
Price (USD)/ Each
75.65
Online exclusive
84.40 
Save 8.75 (10%)
Add to cart
Phosphate-buffered saline (PBS, 10X), pH 7.6
Catalog numberJ62692.K2
Price (USD)/ Each
75.65
Online exclusive
84.40 
Save 8.75 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Capacity1 L
Recommended StorageAmbient temperatures
Dilution10 X
Product TypePhosphate Buffered Saline (PBS)
pH7.6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only