Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals
Gelatin, 10% soln., Thermo Scientific Chemicals
Catalog number: J62699.AK
250 mL, Each
Thermo Scientific Chemicals
Gelatin, 10% soln., Thermo Scientific Chemicals
Catalog number: J62699.AK
250 mL, Each
Quantity
Catalog number: J62699.AK
also known as J62699-AK
Price (USD)
Have Questions?
Description
Gelatin is used for preparing slides for histochemistry and light microscope autoradiography. Gelatin has also been added to cell culture plates and improves the attachment of a variety of cell types. Additionally, gelatin can inhibit crystallization in bacteriology studies. In addition it is an excellent diluents for primary and secondary antibodies.