Thermo Scientific Chemicals

Mefenamic acid, 98%, Thermo Scientific Chemicals

Catalog number: J62705.22
100 g, Each
Thermo Scientific Chemicals

Mefenamic acid, 98%, Thermo Scientific Chemicals

Catalog number: J62705.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62705.22
also known as J62705-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61-68-7
IUPAC Name
2-[(2,3-dimethylphenyl)amino]benzoic acid
Molecular Formula
C15H15NO2
InChI Key
HYYBABOKPJLUIN-UHFFFAOYSA-N
SMILES
CC1=CC=CC(NC2=CC=CC=C2C(O)=O)=C1C
Appearance (Color)
White to pale yellow
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥97.5% (titration)

Description

Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.

Solubility
It is soluble in acetone, chloroform, dichloromethane, methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text