Thermo Scientific Chemicals

FURA 2-AM, Thermo Scientific Chemicals

Catalog number: J62728.MCR
1 mg, Each
Thermo Scientific Chemicals

FURA 2-AM, Thermo Scientific Chemicals

Catalog number: J62728.MCR
1 mg, Each
Quantity
Catalog number: J62728.MCR
also known as J62728-M^
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108964-32-5
IUPAC Name
(acetyloxy)methyl 2-({2-[(acetyloxy)methoxy]-2-oxoethyl}[2-(5-{[(acetyloxy)methoxy]carbonyl}-1,3-oxazol-2-yl)-4-(2-{2-[bis({2-[(acetyloxy)methoxy]-2-oxoethyl})amino]-5-methylphenoxy}ethoxy)-1-benzofuran-5-yl]amino)acetate
Molecular Formula
C44H47N3O24
InChI Key
UTBYGIDPBQOHNT-UHFFFAOYSA-N
SMILES
CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)C1=CC=C(C)C=C1OCCOC1=C2C=C(OC2=CC=C1N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)C1=NC=C(O1)C(=O)OCOC(C)=O
Assay from Suppliers CofA
≥94.0% (HPLC)
Form
Powder
Appearance (Color)
Pale cream to yellow

Description

A calcium indicator; derivative of Fura-2 used to load the chelator into cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A calcium indicator; derivative of Fura-2 used to load the chelator into cells
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text