Thermo Scientific Chemicals

Bupivacaine, Thermo Scientific Chemicals

Catalog number: J62742.14
25 g, Each
Thermo Scientific Chemicals

Bupivacaine, Thermo Scientific Chemicals

Catalog number: J62742.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62742.14
also known as J62742-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38396-39-3
Form
Powder or crystals or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to off-white
Assay from Supplier's CofA
≥97.0%
Loss on Drying
≤0.5%

Description

Bupivacaine acts as a potent and long-acting local anesthetic agent in vivo. At 100 μM, it has been reported to inhibit the uptake of dopamine in striatal synaptosomes by 47%.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Application

  • Bupivacaine has been shown to activate the AMPKα/BDNF signaling pathway in spinal astrocytes
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text