Orange G, Electrophoresis Grade, Thermo Scientific Chemicals
Orange G, Electrophoresis Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Orange G, Electrophoresis Grade, Thermo Scientific Chemicals

Collagen stain for connective tissue
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
1 kg
Catalog number J62743.22
also known as J62743-22
Price (USD)/ Each
40.50
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
40.50
Add to cart
Orange G, Electrophoresis Grade, Thermo Scientific Chemicals
Catalog numberJ62743.22
Price (USD)/ Each
40.50
-
Add to cart
Chemical Identifiers
CAS1936-15-8
IUPAC Namedisodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
Molecular FormulaC16H10N2Na2O7S2
InChI KeyAEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying10.0% max.
Assay (unspecified)Dye content: 90% min.
Assay (unspecified)Lambda max: 479nm
Collagen stain for connective tissue

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only