Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals
Gelatin Blocking Buffer, 1% in PBS
Catalog number: J62755.K2
1 L, Each
Thermo Scientific Chemicals
Gelatin Blocking Buffer, 1% in PBS
Catalog number: J62755.K2
1 L, Each
Quantity
Catalog number: J62755.K2
also known as J62755-K2
Price (USD)
Have Questions?
Description
Suitable for use as a non-specific blocking agent for Western blots and ELISA assays. It is used to block non-specific binding on the membrane, when using nylon or nitrocellulose (but not PVDF).