Evodiamine
Thermo Scientific Chemicals

Evodiamine

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62771.MD
also known as J62771-MD
Price (USD)/ Each
124.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
124.00
Add to cart
Evodiamine
Catalog numberJ62771.MD
Price (USD)/ Each
124.00
-
Add to cart
Specifications
Quantity250 mg
DescriptionOff-white to pale yellow
For Use With (Application)A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells
Recommended StorageStore at -20°C
CAS518-17-2
View more
A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C