TRIS-buffered saline (TBS, 10X) pH 8.0
TRIS-buffered saline (TBS, 10X) pH 8.0
Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 10X) pH 8.0

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2 L
4 L
Catalog number J62780.K2
also known as J62780-K2
Price (USD)/ Each
61.65
Online exclusive
68.60 
Save 6.95 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
68.60 
Save 6.95 (10%)
Add to cart
TRIS-buffered saline (TBS, 10X) pH 8.0
Catalog numberJ62780.K2
Price (USD)/ Each
61.65
Online exclusive
68.60 
Save 6.95 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialTRIS-buffered saline
CAS77-86-1
Recommended StorageAmbient temperatures
Concentration10X
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only