Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, 0.5M aq. soln, pH 8.0

Catalog number: J62786.AK
250 mL, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, 0.5M aq. soln, pH 8.0

Catalog number: J62786.AK
250 mL, Each
Quantity
Catalog number: J62786.AK
also known as J62786-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-00-4
IUPAC Name
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Molecular Formula
C10H16N2O8
InChI Key
KCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILES
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Appearance (Color)
Clear colorless
Form
Liquid
Comment
All components are dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment, 0.22 micron filter.
pH
8.0 ± 0.2

Description

Ethylenediaminetetraacetic acid solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, chelators and ethylenediaminetetraacetic acid (EDTA) and EDTA solutions. Ethylenediaminetetraacetic acid solution has been used in a study to help elucidate the role of antigen retrieval in immunostaining of ethanol-fixed smears. Ethylenediaminetetraacetic acid solution has also been used in a study to assess the uptake and release of biotin-labeled fluorescein (b-FITC) as well as immobilization of biotin-labeled glucose oxidase (b-GOx) to the polyelectrolyte microcapsules with an avidin-poly(styrene sulfonate) (PSS) membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, chelators and ethylenediaminetetraacetic acid (EDTA) and EDTA solutions. Ethylenediaminetetraacetic acid solution has been used in a study to help elucidate the role of antigen retrieval in immunostaining of ethanol-fixed smears. Ethylenediaminetetraacetic acid solution has also been used in a study to assess the uptake and release of biotin-labeled fluorescein (b-FITC) as well as immobilization of biotin-labeled glucose oxidase (b-GOx) to the polyelectrolyte microcapsules with an avidin-poly(styrene sulfonate) (PSS) membrane.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text