Thermo Scientific Chemicals

Dextran sulfate sodium salt, MW ca >500,000, Thermo Scientific Chemicals

Catalog number: J62787.18
50 g, Each
Thermo Scientific Chemicals

Dextran sulfate sodium salt, MW ca >500,000, Thermo Scientific Chemicals

Catalog number: J62787.18
50 g, Each
Quantity
Catalog number: J62787.18
also known as J62787-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9011-18-1
IUPAC Name
Dextran sulfate sodium salt
MDL Number
MFCD00081551
Appearance (Color)
White to cream to pale yellow
Form
Powder
Assay (unspecified)
Specific viscosity: 0.10 to 0.70
Assay (special)
Total Sulfur content: 17.0 to 20.0%
Chloride content
≤1000 ppm

Description

Dextran sulfate sodium is used to study colitis (inflammatory bowel disease) as well as an antiviral agent. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text