Thermo Scientific Chemicals

TBE running buffer (10X)

Catalog number: J62788.K7
4 L, Each
Thermo Scientific Chemicals

TBE running buffer (10X)

Catalog number: J62788.K7
4 L, Each
Quantity
Catalog number: J62788.K7
also known as J62788-K7
Price (USD)
Quantity
-
Have Questions?
Form
Liquid
Appearance (Color)
Clear colorless
Comment
All components are dissolved in UV treated 18.2 megohm-cm-1 water, followed by pH adjustment and 1 micron filtration
Composition
0.89M Tris, 0.89 boric acid, 20mM EDTA
pH
8.0-8.5

Description

TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. TBE Buffer is used for polyacrylamide and agarose gel electrophoresis. It is ideally suited for DNA and RNA separation on longer run, higher voltage or amperage gels. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel.5 TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids. Tris-Borate-EDTA buffer has been used for pulsed-field gel electrophoresis (PFGE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text