Thermo Scientific Chemicals

Vancomycin hydrochloride, Molecular Biology Grade

Catalog number: J62790.03
1 g, Each
Thermo Scientific Chemicals

Vancomycin hydrochloride, Molecular Biology Grade

Catalog number: J62790.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62790.03
also known as J62790-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1404-93-9
IUPAC Name
48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
Molecular Formula
C66H76Cl3N9O24
InChI Key
LCTORFDMHNKUSG-UHFFFAOYNA-N
SMILES
Cl.CNC(CC(C)C)C(=O)NC1C(O)C2=CC(Cl)=C(OC3=CC4=CC(OC5=C(Cl)C=C(C=C5)C(O)C5NC(=O)C(NC(=O)C4NC(=O)C(CC(N)=O)NC1=O)C1=CC(=C(O)C=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C=C2
Appearance (Color)
White to tan to brown
Form
Free-flowing powder
Assay (HPLC)
Vancomycin B: ≥85.0%
Heavy Metals
≤30 ppm
Water Content (Karl Fischer Titration)
≤5.0%

Description

Inhibits bacterial mucopeptide synthesis.Vancomycin hydrochloride is used as an antiviral and anti bacterial. binds to bacterial cell wall. It exhibits potent activity against Gram positive bacteria and is highly effective against MRSA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vancomycin hydrochloride is a glycopeptide antibiotic that prevents cell wall synthesis
  • Originally obtained from the bacterial strain Streptomyces Orientalis
  • Vancomycin binds to D-alanyl-D-alanine motifs in cell wall precursors and leads to cell death
  • It is active against some penicillin-resistant bacteria

Application

  • Vancomycin hydrochloride is intended for use in the research of infections and for antibiotic resistance testing
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text