Deblock trichloroacetic acid soln., 3% w/w in dichloromethane, Thermo Scientific Chemicals
Deblock trichloroacetic acid soln., 3% w/w in dichloromethane, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Deblock trichloroacetic acid soln., 3% w/w in dichloromethane, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
450 mL
2.5 L
Catalog number J62821.AO
also known as J62821-AO
Price (USD)/ Each
71.40
-
Add to cart
Quantity:
450 mL
Request bulk or custom format
Price (USD)/ Each
71.40
Add to cart
Deblock trichloroacetic acid soln., 3% w/w in dichloromethane, Thermo Scientific Chemicals
Catalog numberJ62821.AO
Price (USD)/ Each
71.40
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialDeblock trichloroacetic acid soln.
ColorColorless
Health Hazard 1H315-H318-H336-H350
Health Hazard 2GHS H Statement
H318-H351-H315
Causes serious eye damage.
Suspected of causing cancer.
Causes skin irritation.
Health Hazard 3P201-P202-P261-P264b-P271-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P332+P313-P362-P501c
View more
Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only