Thermo Scientific Chemicals

Deblock trichloroacetic acid soln., 3% w/w in dichloromethane, Thermo Scientific Chemicals

Catalog number: J62821.K4
2.5 L, Each
Thermo Scientific Chemicals

Deblock trichloroacetic acid soln., 3% w/w in dichloromethane, Thermo Scientific Chemicals

Catalog number: J62821.K4
2.5 L, Each
Quantity
Catalog number: J62821.K4
also known as J62821-K4
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
Clear, colorless
Form
Liquid
Water Content (Karl Fischer Titration)
150ppm max.

Description

Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text