Tween 20 washing buffer, 1.0% in PBS (20X)
Tween 20 washing buffer, 1.0% in PBS (20X)
Thermo Scientific Chemicals

Tween 20 washing buffer, 1.0% in PBS (20X)

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2 L
Catalog number J62844.K2
also known as J62844-K2
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
Add to cart
Tween 20 washing buffer, 1.0% in PBS (20X)
Catalog numberJ62844.K2
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Specifications
Product TypeBuffer
Quantity1 L
FormLiquid
Concentration1.0% in PBS (20X)
Unit SizeEach
It is used as wash buffers for ELISA, Western blot and other immunoassays as well as a blocking buffer for plate-based assays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
Ambient temperatures