Thermo Scientific Chemicals

Phosphate-buffered saline (PBS, 10X), RNase free

Catalog number: J62851.AP
500 mL, Each
Thermo Scientific Chemicals

Phosphate-buffered saline (PBS, 10X), RNase free

Catalog number: J62851.AP
500 mL, Each
Capacity
Catalog number: J62851.AP
also known as J62851-AP
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
Clear colorless
Comment
All components are dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and filtered through 0.22 micron filter.
pH
7.4 ± 0.2
Form
Liquid

Description

Phosphate-buffered saline (PBS, 10X), is an isotonic, non-toxic to most cells and is used in biological research. Phosphate-buffered saline with EDTA is used to disengage attached and clumped cells. It is also used in cell culturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text