Ethyl Orange sodium salt, Thermo Scientific Chemicals
Ethyl Orange sodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl Orange sodium salt, Thermo Scientific Chemicals

Indicator: pH 3.0 (red) to pH 4.5 (yellow)
Have Questions?
Quantity:
100 g
Catalog number J62863.22
also known as J62863-22
Price (USD)/ Each
35.50
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
35.50
Add to cart
Ethyl Orange sodium salt, Thermo Scientific Chemicals
Catalog numberJ62863.22
Price (USD)/ Each
35.50
-
Add to cart
Specifications
Chemical Name or MaterialEthyl Orange sodium salt
CAS62758-12-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Indicator: pH 3.0 (red) to pH 4.5 (yellow)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only