Thermo Scientific Chemicals

Thapsigargin, 95%, Thermo Scientific Chemicals

Catalog number: J62866.MA
10 mg, Each
Thermo Scientific Chemicals

Thapsigargin, 95%, Thermo Scientific Chemicals

Catalog number: J62866.MA
10 mg, Each
Quantity
Catalog number: J62866.MA
also known as J62866-MA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67526-95-8
IUPAC Name
(3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-(acetyloxy)-4-(butanoyloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-{[(2Z)-2-methylbut-2-enoyl]oxy}-2-oxo-2H,3H,3aH,4H,5H,6H,6aH,7H,8H,9bH-azuleno[4,5-b]furan-7-yl octanoate
Molecular Formula
C34H50O12
InChI Key
IXFPJGBNCFXKPI-FSIHEZPISA-N
SMILES
CCCCCCCC(=O)O[C@@H]1[C@@H](OC(=O)C(\C)=C/C)C(C)=C2[C@@H]3OC(=O)[C@@](C)(O)[C@@]3(O)[C@H](C[C@](C)(OC(C)=O)[C@@H]12)OC(=O)CCC
Form
Wax/film, lyophilized
Assay from Supplier's CofA
≥94.0%

Description

Thapsigargin- induces down regulation of the EGF receptor is independent of protein kinase C activation. It is also used as an inducer of stored Ca2+ release.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thapsigargin- induces down regulation of the EGF receptor is independent of protein kinase C activation. It is also used as an inducer of stored Ca2+ release.

Solubility
Soluble in DMSO (20 mg/ml), ethanol (20 mg/ml), and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text