L-Methionine sulfoxide
L-Methionine sulfoxide
Thermo Scientific Chemicals

L-Methionine sulfoxide

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number J62873.03
also known as J62873-03
Price (USD)/ Each
41.65
Online exclusive
46.00 
Save 4.35 (9%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
46.00 
Save 4.35 (9%)
Add to cart
L-Methionine sulfoxide
Catalog numberJ62873.03
Price (USD)/ Each
41.65
Online exclusive
46.00 
Save 4.35 (9%)
-
Add to cart
Chemical Identifiers
CAS3226-65-1
IUPAC Name(2S)-2-amino-4-methanesulfinylbutanoic acid
Molecular FormulaC5H11NO3S
InChI KeyQEFRNWWLZKMPFJ-YGVKFDHGSA-N
SMILESCS(=O)CC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point236 - 248 °C
Appearance (Color)White to off-white
Assay (HPLC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only