Thermo Scientific Chemicals

Aztreonam, Thermo Scientific Chemicals

Catalog number: J62887.06
5 g, Each
Thermo Scientific Chemicals

Aztreonam, Thermo Scientific Chemicals

Catalog number: J62887.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62887.06
also known as J62887-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78110-38-0
IUPAC Name
(2S,3S)-3-[(2Z)-2-(2-azaniumyl-1,3-thiazol-4-yl)-2-[(1-carboxy-1-methylethoxy)imino]acetamido]-2-methyl-4-oxoazetidine-1-sulfonate
Molecular Formula
C13H17N5O8S2
InChI Key
WZPBZJONDBGPKJ-VEHQQRBSSA-N
SMILES
C[C@H]1[C@H](NC(=O)C(=N/OC(C)(C)C(O)=O)\C2=CSC([NH3+])=N2)C(=O)N1S([O-])(=O)=O
Comment
Related substances: ≤ 1.5% Individual, ≤ 2.5 % Total
Water Content (Karl Fischer Titration)
≤2%
Assay (HPLC)
≥92.0%
Heavy Metals
≤10 ppm
Optical Rotation
- 26° to -32°

Description

Aztreonam is used to treat severe infections of the urinary tract, lower respiratory tract, skin, stomach, female reproductive organs, and other body systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aztreonam is used to treat severe infections of the urinary tract, lower respiratory tract, skin, stomach, female reproductive organs, and other body systems.

Solubility
Soluble in DMF/water (1:1) at 50 mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text