Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals
Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J62905.AP
also known as J62905-AP
Price (USD)/ Each
88.65
Online exclusive
97.80 
Save 9.15 (9%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
88.65
Online exclusive
97.80 
Save 9.15 (9%)
Add to cart
Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals
Catalog numberJ62905.AP
Price (USD)/ Each
88.65
Online exclusive
97.80 
Save 9.15 (9%)
-
Add to cart
Specifications
Chemical Name or MaterialCalcium chloride
Name Notesterile
CAS10043-52-4
Recommended StorageKeep cold
Physical FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only