Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals
Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number J62905.K2
also known as J62905-K2
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
Add to cart
Calcium chloride, 100mM aq. soln., sterile-filtered, Thermo Scientific Chemicals
Catalog numberJ62905.K2
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialCalcium chloride
Name Notesterile
CAS10043-52-4
Recommended StorageKeep cold
Physical FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only